APOH11A9E008 Apollo Scientific Ltd

Ethyl (4-oxocyclohex-1-yl)acetate

  • MDL number MFCD11045783
  • Cas Number : 58012-34-3
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (4-Oxocyclohex-1-yl)acetic acid ethyl ester, Ethyl 2-(4-oxocyclohex-1-yl)ethanoate
Purity 95
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-4-oxocyclohex-1-ylacetate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-4-oxocyclohex-1-ylacetate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart