APOH11A9E0B6 Apollo Scientific Ltd

2,4-Dibromobenzaldehyde

  • MDL number MFCD13185489
  • Cas Number : 5629-98-1
  • Molecular Weight : 263.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 80
Iupac Name 2,4-dibromobenzaldehyde
Inchi Code 1S/C7H4Br2O/c8-6-2-1-5(4-10)7(9)3-6/h1-4H
Inchi Key PGSVNTIDEZRNES-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 80
Product Link https://store.apolloscientific.co.uk/product/24-dibromobenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-dibromobenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart