APOH11A9E0DC Apollo Scientific Ltd

Methyl 2-hydroxy-4-nitrobenzoate

  • MDL number MFCD11041295
  • Cas Number : 13684-28-1
  • Molecular Weight : 197.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 101-102
Iupac Name methyl 2-hydroxy-4-nitrobenzoate
Inchi Code 1S/C8H7NO5/c1-14-8(11)6-3-2-5(9(12)13)4-7(6)10/h2-4,10H,1H3
Inchi Key DODUOCYKSQVFSW-UHFFFAOYSA-N

Description

Synonym Methyl 4-nitrosalicylate, 3-Hydroxy-4-(methoxycarbonyl)nitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 101-102
Product Link https://store.apolloscientific.co.uk/product/methyl-2-hydroxy-4-nitrobenzoate

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/methyl-2-hydroxy-4-nitrobenzoate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart