APOH11A9E22C Apollo Scientific Ltd

Ethyl 5-bromothiophene-3-carboxylate

  • MDL number MFCD14155650
  • Cas Number : 170355-38-1
  • Molecular Weight : 235.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-bromothiophene-3-carboxylate
Inchi Code 1S/C7H7BrO2S/c1-2-10-7(9)5-3-6(8)11-4-5/h3-4H,2H2,1H3
Inchi Key LUYMKCLOYODOEI-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-5-bromothiophene-3-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-5-bromothiophene-3-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart