APOH11A9E293 Apollo Scientific Ltd

3-Bromo-6-chloro-2-methoxypyridine

  • MDL number MFCD18257579
  • Cas Number : 1211526-62-3
  • Molecular Weight : 222.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-6-chloro-2-methoxypyridine

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/3-bromo-6-chloro-2-methoxypyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart