APOH11A9E4C7 Apollo Scientific Ltd

4-Chloro-2-iodobenzenesulfonyl chloride

  • MDL number MFCD18393342
  • Cas Number : 1261730-58-8
  • Molecular Weight : 336.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Chloro-2-iodobenzenesulfonyl chloride
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-2-iodobenzenesulfonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-2-iodobenzenesulfonyl-chloride
pictograms GHS05;GHS06
hstatements H301+H311+H331;H314
Availability details for
Quantity
  • Availability
  • Add to Cart