APOH11A9E4FF Apollo Scientific Ltd

2-Amino-6-nitrobenzoic acid

  • MDL number MFCD00034785
  • Cas Number : 50573-74-5
  • Molecular Weight : 182.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-6-nitrobenzoic acid
Inchi Code 1S/C7H6N2O4/c8-4-2-1-3-5(9(12)13)6(4)7(10)11/h1-3H,8H2,(H,10,11)
Inchi Key GGKYLHNARFFORH-UHFFFAOYSA-N

Description

Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-6-nitrobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-6-nitrobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart