APOH11A9E50C Apollo Scientific Ltd

Thiomorpholine, N-BOC protected

  • MDL number MFCD14635788
  • Cas Number : 220655-09-4
  • Molecular Weight : 203.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-thiomorpholinecarboxylate
Inchi Code 1S/C9H17NO2S/c1-9(2,3)12-8(11)10-4-6-13-7-5-10/h4-7H2,1-3H3
Inchi Key CTJYSVRXJDACIO-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/thiomorpholine-n-boc-protected

Safety Information

signalword Danger
pcodes P260;P280;P301+P330+P331
MSDS https://store.apolloscientific.co.uk/product/thiomorpholine-n-boc-protected
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart