APOH11A9E50E Apollo Scientific Ltd

2-Amino-6-nitrobenzyl alcohol

  • MDL number MFCD01664411
  • Cas Number : 98451-51-5
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-amino-6-nitrophenyl)methanol
Inchi Code 1S/C7H8N2O3/c8-6-2-1-3-7(9(11)12)5(6)4-10/h1-3,10H,4,8H2
Inchi Key BSQXKAWQRQDMAK-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-6-nitrobenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-6-nitrobenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart