APOH11A9E50F Apollo Scientific Ltd

Methyl 2-amino-6-nitrobenzoate

  • MDL number MFCD01851258
  • Cas Number : 57113-89-0
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-amino-6-nitrobenzoate
Inchi Code 1S/C8H8N2O4/c1-14-8(11)7-5(9)3-2-4-6(7)10(12)13/h2-4H,9H2,1H3
Inchi Key NFPMHGVGDWXWRJ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-amino-6-nitrobenzoate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-amino-6-nitrobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart