APOH11A9E51D Apollo Scientific Ltd

5-Chloro-2-hydroxyphenylacetic acid

  • MDL number MFCD11840335
  • Cas Number : 24161-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(5-Chloro-2-hydroxyphenyl)acetic acid
Purity 95
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=117832

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=117832
Availability details for
Quantity
  • Availability
  • Add to Cart