APOH11C5ADC6 Apollo Scientific Ltd

(4-Aminothiophen-2-yl)boronic acid hydrochloride

  • MDL number MFCD28670063

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=118285

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340;P305+P351+P338;P321;P362+P364
MSDS http://www.apolloscientific.co.uk/display_item.php?id=118285
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart