APOH11C5ADEA Apollo Scientific Ltd

1-(Cyclopenta-2,4-dien-1-ylidene)ethyl acetate

  • MDL number MFCD30749182
  • Cas Number : 701-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 93
Storage temp. Ambient temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=118297

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=118297
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart