APOH11C5ADFF Apollo Scientific Ltd

2-(Piperidin-3-yl)ethan-1-ol hydrochloride

  • MDL number MFCD14583117
  • Cas Number : 16780-04-4
  • Molecular Weight : 165.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-piperidinyl)ethanol hydrochloride
Inchi Code 1S/C7H15NO.ClH/c9-5-3-7-2-1-4-8-6-7;/h7-9H,1-6H2;1H
Inchi Key JPWLRMLPKPPHRJ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-piperidin-3-ylethan-1-ol-hydrochloride

Safety Information

signalword Warning
pcodes P280;P301+P312;P302+P352
MSDS https://store.apolloscientific.co.uk/product/2-piperidin-3-ylethan-1-ol-hydrochloride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart