APOH11C5AE03 Apollo Scientific Ltd

tert-Butyl 3-cyanopiperazine-1-carboxylate

  • MDL number MFCD09991479
  • Cas Number : 859518-35-7
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-cyano-1-piperazinecarboxylate
Inchi Code 1S/C10H17N3O2/c1-10(2,3)15-9(14)13-5-4-12-8(6-11)7-13/h8,12H,4-5,7H2,1-3H3
Inchi Key CBFXRUGJRMBDFG-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/tert-butyl-3-cyanopiperazine-1-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/tert-butyl-3-cyanopiperazine-1-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart