APOH11C5AE08 Apollo Scientific Ltd

tert-Butyl (2-(pyrrolidin-3-yl)ethyl)carbamate

  • MDL number MFCD12828830
  • Cas Number : 169750-93-0
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-(3-pyrrolidinyl)ethylcarbamate
Inchi Code 1S/C11H22N2O2/c1-11(2,3)15-10(14)13-7-5-9-4-6-12-8-9/h9,12H,4-8H2,1-3H3,(H,13,14)
Inchi Key OWGYHEOEGOLPEK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/tert-butyl-2-pyrrolidin-3-ylethylcarbamate

Safety Information

signalword Warning
pcodes P304+P340
MSDS https://store.apolloscientific.co.uk/product/tert-butyl-2-pyrrolidin-3-ylethylcarbamate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart