APOH11C5AE13 Apollo Scientific Ltd

5-Bromo-1-methyl-1H-indole-3-carbaldehyde

  • MDL number MFCD03906373
  • Cas Number : 10102-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-135
Purity 97
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=118353

Safety Information

signalword Warning
pcodes P280;P301+P312;P302+P352
MSDS http://www.apolloscientific.co.uk/display_item.php?id=118353
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart