APOH11C5AE17 Apollo Scientific Ltd

4,6-Dimethoxy-5-methyl-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD28165860
  • Cas Number : 57122-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=118357

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=118357
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart