APOH11C5AEAA Apollo Scientific Ltd

1-Benzyl-3-bromopyrrolidine

  • MDL number MFCD11226925
  • Cas Number : 38042-74-9
  • Molecular Weight : 240.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-benzyl-3-bromopyrrolidine

Safety Information

MSDS https://store.apolloscientific.co.uk/product/1-benzyl-3-bromopyrrolidine
Availability details for
Quantity
  • Availability
  • Add to Cart