APOH11C5AF20 Apollo Scientific Ltd

(S)-2-Chloro-1-(3,4-difluorophenyl)ethanol

  • MDL number MFCD09863590
  • Cas Number : 1006376-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/s-2-chloro-1-34-difluorophenylethanol-99

Safety Information

signalword Danger
pcodes P280;P301+P312;P302+P352
MSDS https://store.apolloscientific.co.uk/product/s-2-chloro-1-34-difluorophenylethanol-99
pictograms GHS05;GHS07
hstatements H302+H312+H332;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart