APOH11C5AF82 Apollo Scientific Ltd

1-(3,4-Difluorophenyl)ethanol

  • MDL number MFCD08442288
  • Cas Number : 321318-21-2
  • Molecular Weight : 158.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,4-difluorophenyl)ethanol
Inchi Code 1S/C8H8F2O/c1-5(11)6-2-3-7(9)8(10)4-6/h2-5,11H,1H3
Inchi Key WSIJRUFDDILTTN-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-34-difluorophenylethanol

Safety Information

signalword Warning
pcodes P280;P271;P261
MSDS https://store.apolloscientific.co.uk/product/1-34-difluorophenylethanol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart