APOH11DCF62F Apollo Scientific Ltd

2-Hydroxy-n-butyric acid ethyl ester

  • MDL number MFCD00014407
  • Cas Number : 52089-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=119774

Safety Information

signalword Warning
pcodes P280;P302+P352;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=119774
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart