APOH14DBAA10 Apollo Scientific Ltd

tert-Butyl (R)-(1-((trifluoromethyl)thio)propan-2-yl)carbamate

  • MDL number MFCD31653055

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=120323

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=120323
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart