APOH14DBAA11 Apollo Scientific Ltd

tert-Butyl (3-((trifluoromethyl)thio)propyl)carbamate

  • MDL number MFCD17677143
  • Cas Number : 1208080-46-9
  • Molecular Weight : 259.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-[(trifluoromethyl)sulfanyl]propylcarbamate
Inchi Code 1S/C9H16F3NO2S/c1-8(2,3)15-7(14)13-5-4-6-16-9(10,11)12/h4-6H2,1-3H3,(H,13,14)
Inchi Key SYQRXTZVLYOPJT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/tert-butyl-3-trifluoromethylthiopropylcarbamate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/tert-butyl-3-trifluoromethylthiopropylcarbamate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart