APOH14DBAA6C Apollo Scientific Ltd

Butyl 2,2-difluorocyclopropanecarboxylate

  • MDL number MFCD06797747
  • Cas Number : 260352-79-2
  • Molecular Weight : 178.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name butyl 2,2-difluorocyclopropanecarboxylate
Inchi Code 1S/C8H12F2O2/c1-2-3-4-12-7(11)6-5-8(6,9)10/h6H,2-5H2,1H3
Inchi Key JHIWVOJDXOSYLW-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/butyl-22-difluorocyclopropanecarboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/butyl-22-difluorocyclopropanecarboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart