APOH14DBAA98 Apollo Scientific Ltd

3-(Difluoromethyl)-2,6-difluoropyridine

  • MDL number MFCD22199312
  • Cas Number : 85396-63-0
  • Molecular Weight : 165.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-difluoromethyl-26-difluoropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-difluoromethyl-26-difluoropyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart