APOH14DBAABF Apollo Scientific Ltd

1-Fluoro-4-(pentafluoroethyl)benzene

  • MDL number MFCD16140153
  • Cas Number : 2396-11-4
  • Molecular Weight : 214.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-fluoro-4-pentafluoroethylbenzene

Safety Information

MSDS https://store.apolloscientific.co.uk/product/1-fluoro-4-pentafluoroethylbenzene
Availability details for
Quantity
  • Availability
  • Add to Cart