APOH14DBAB13 Apollo Scientific Ltd

2-[3-Bromo-5-(trifluoromethyl)phenyl]acetic acid

  • MDL number MFCD13194218
  • Cas Number : 1161362-01-1
  • Molecular Weight : 283.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-bromo-5-(trifluoromethyl)phenyl]acetic acid
Inchi Code 1S/C9H6BrF3O2/c10-7-2-5(3-8(14)15)1-6(4-7)9(11,12)13/h1-2,4H,3H2,(H,14,15)
Inchi Key POLZAYSCKXDHKM-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-3-bromo-5-trifluoromethylphenylacetic-acid

Safety Information

signalword Danger
pcodes P301+P312;P301+P330+P331
MSDS https://store.apolloscientific.co.uk/product/2-3-bromo-5-trifluoromethylphenylacetic-acid
pictograms GHS05;GHS07
hstatements H302;H314;H317
Availability details for
Quantity
  • Availability
  • Add to Cart