APOH14DBABCA Apollo Scientific Ltd

2,3,4-Trifluoro-6-(trifluoromethyl)benzaldehyde

  • MDL number MFCD31634834
  • Cas Number : 2244084-38-4
  • Molecular Weight : 228.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3,4-trifluoro-6-(trifluoromethyl)benzaldehyde
Inchi Code 1S/C8H2F6O/c9-5-1-4(8(12,13)14)3(2-15)6(10)7(5)11/h1-2H
Inchi Key UVWRLIFRSOYKRQ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/234-trifluoro-6-trifluoromethylbenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/234-trifluoro-6-trifluoromethylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart