APOH14DBABCF Apollo Scientific Ltd

5-Bromo-2-(trifluoromethyl)benzyl fluoride

  • MDL number MFCD31634841

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=120452

Safety Information

signalword Danger
pcodes P301+P330+P331;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=120452
pictograms GHS05;GHS08
hstatements H314;H351
Availability details for
Quantity
  • Availability
  • Add to Cart