APOH14DBABF4 Apollo Scientific Ltd

4-Cyclopropylbenzenesulphonyl chloride

  • MDL number MFCD09929602
  • Cas Number : 167404-32-2
  • Molecular Weight : 216.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-cyclopropylbenzenesulfonyl chloride
Inchi Code 1S/C9H9ClO2S/c10-13(11,12)9-5-3-8(4-6-9)7-1-2-7/h3-7H,1-2H2
Inchi Key QCHUBTJEGZWBFJ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-cyclopropylbenzenesulphonyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-cyclopropylbenzenesulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart