APOH14DBAC5D Apollo Scientific Ltd

4-Bromo-2-methylbenzyl fluoride

  • MDL number MFCD22492378
  • Cas Number : 1824142-81-5
  • Molecular Weight : 203.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-1-(fluoromethyl)-2-methylbenzene
Inchi Code 1S/C8H8BrF/c1-6-4-8(9)3-2-7(6)5-10/h2-4H,5H2,1H3
Inchi Key KREHZAOZXBYTEJ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-methylbenzyl-fluoride

Safety Information

signalword Danger
pcodes P260;P280;P301+P330+P331
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-methylbenzyl-fluoride
pictograms GHS05;GHS08
hstatements H314;H351
Availability details for
Quantity
  • Availability
  • Add to Cart