APOH29F2C46C Apollo Scientific Ltd

4-Ethynyltetrahydro-2H-pyran

  • MDL number MFCD17016081
  • Cas Number : 1202245-65-5
  • Molecular Weight : 110.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-ethynyltetrahydro-2h-pyran

Safety Information

signalword Danger
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/4-ethynyltetrahydro-2h-pyran
pictograms GHS02;GHS07
hstatements H225;H315;H319;H335;H336
Availability details for
Quantity
  • Availability
  • Add to Cart