APOH2F622739 Apollo Scientific Ltd

1-Methyl-7-nitroisatoic anhydride

  • MDL number MFCD18432729
  • Cas Number : 73043-80-8
  • Molecular Weight : 222.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 203-206
Iupac Name 1-methyl-7-nitro-2H-3,1-benzoxazine-2,4(1H)-dione
Inchi Code 1S/C9H6N2O5/c1-10-7-4-5(11(14)15)2-3-6(7)8(12)16-9(10)13/h2-4H,1H3
Inchi Key MULNCJWAVSDEKJ-UHFFFAOYSA-N

Description

Synonym 1-Methyl-7-nitro-3,1-benzoxazine-2,4-dione
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 203-206
Product Link https://store.apolloscientific.co.uk/product/1-methyl-7-nitroisatoic-anhydride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-methyl-7-nitroisatoic-anhydride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart