APOH2F62273B Apollo Scientific Ltd

4-Chloro-5-methyl-1H-indazole

  • MDL number MFCD22628251
  • Cas Number : 152626-83-0
  • Molecular Weight : 166.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-5-methyl-1H-indazole
Inchi Code 1S/C8H7ClN2/c1-5-2-3-7-6(8(5)9)4-10-11-7/h2-4H,1H3,(H,10,11)
Inchi Key RHXGTDAHAAPERW-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-5-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/4-chloro-5-methyl-1h-indazole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart