APOH2F62273D Apollo Scientific Ltd

4-Bromo-5-chloro-1H-indazole

  • MDL number MFCD12828060
  • Cas Number : 1056264-74-4
  • Molecular Weight : 231.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-5-chloro-1H-indazole
Inchi Code 1S/C7H4BrClN2/c8-7-4-3-10-11-6(4)2-1-5(7)9/h1-3H,(H,10,11)
Inchi Key HBWQXAIHQCZDAB-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-5-chloro-1h-indazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-5-chloro-1h-indazole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart