APOH2F622745 Apollo Scientific Ltd

(2S)-2-Trifluoromethylpyrrolidine hydrochloride

  • MDL number MFCD22380026
  • Cas Number : 1410903-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=121412

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=121412
pictograms GHS02;GHS07
hstatements H228;H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart