APOH2F622746 Apollo Scientific Ltd

4-Bromo-2-fluoro-5-methylphenol

  • MDL number MFCD09261252
  • Cas Number : 550400-07-2
  • Molecular Weight : 205.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 60-62
Iupac Name 4-bromo-2-fluoro-5-methylphenol
Inchi Code 1S/C7H6BrFO/c1-4-2-7(10)6(9)3-5(4)8/h2-3,10H,1H3
Inchi Key QDGUFFYQTIQGAJ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 60-62
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-fluoro-5-methylphenol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-fluoro-5-methylphenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart