APOH30347620 Apollo Scientific Ltd

1-(3-Bromophenyl)-N-methylethanamine hydrochloride

  • MDL number MFCD30609564
  • Cas Number : 2197556-55-9
  • Molecular Weight : 250.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-bromophenyl)-N-methylethan-1-amine hydrochloride
Inchi Code 1S/C9H12BrN.ClH/c1-7(11-2)8-4-3-5-9(10)6-8;/h3-7,11H,1-2H3;1H
Inchi Key FYUHQQUZYCRREC-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-3-bromophenyl-n-methylethanamine-hydrochloride

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/1-3-bromophenyl-n-methylethanamine-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart