APOH30347723 Apollo Scientific Ltd

Tetrabromo-1,4-benzoquinone

  • MDL number MFCD00013785
  • Cas Number : 488-48-2
  • Molecular Weight : 423.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3,5,6-tetrabromobenzo-1,4-quinone
Inchi Code 1S/C6Br4O2/c7-1-2(8)6(12)4(10)3(9)5(1)11
Inchi Key LWHDQPLUIFIFFT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/tetrabromo-14-benzoquinone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/tetrabromo-14-benzoquinone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart