APOH303479D4 Apollo Scientific Ltd

(2S)-2-Bromo-3-phenylpropanoic acid 1g pack

  • MDL number MFCD00210121
  • Cas Number : 35016-63-8
  • Molecular Weight : 229.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2s-2-bromo-3-phenylpropanoic-acid_OR480663

Safety Information

signalword Warning
pcodes P261;P271;P280
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart