APOH30347AD0 Apollo Scientific Ltd

2-Bromo-1-(4-cyclopropylphenyl)ethanone

  • MDL number MFCD29040961
  • Cas Number : 1934754-00-3
  • Molecular Weight : 239.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-1-(4-cyclopropylphenyl)ethan-1-one
Inchi Code 1S/C11H11BrO/c12-7-11(13)10-5-3-9(4-6-10)8-1-2-8/h3-6,8H,1-2,7H2
Inchi Key YIPSEYWLPHHHND-UHFFFAOYSA-N

Description

Storage temp. Store at -20
Physical Form Liquid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-1-4-cyclopropylphenylethanone

Safety Information

signalword Danger
pcodes P271;P260;P280
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart