APOH30347AD1 Apollo Scientific Ltd

2-Bromo-3'-cyclopropylacetophenone

  • MDL number MFCD20484207
  • Cas Number : 627463-23-4
  • Molecular Weight : 239.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-1-(3-cyclopropylphenyl)ethan-1-one
Inchi Code 1S/C11H11BrO/c12-7-11(13)10-3-1-2-9(6-10)8-4-5-8/h1-3,6,8H,4-5,7H2
Inchi Key AMCOONRENOMFCT-UHFFFAOYSA-N

Description

Storage temp. Store at -20
Physical Form Liquid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-cyclopropylacetophenone

Safety Information

signalword Danger
pcodes P271;P260;P280
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart