APOH30347AD7 Apollo Scientific Ltd

5-Amino-4-bromo-2-chlorophenol

  • MDL number MFCD27942409
  • Cas Number : 1232505-77-9
  • Molecular Weight : 222.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-amino-4-bromo-2-chlorophenol
Inchi Code 1S/C6H5BrClNO/c7-3-1-4(8)6(10)2-5(3)9/h1-2,10H,9H2
Inchi Key AHVAEBUVWFXKIV-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-4-bromo-2-chlorophenol

Safety Information

signalword Warning
pcodes P280;P301+P312
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart