APOH30347AF0 Apollo Scientific Ltd

8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine

  • MDL number MFCD01315052
  • Cas Number : 478066-06-7
  • Molecular Weight : 221.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-chloro-6-trifluoromethyl-124triazolo43-apyridine

Safety Information

signalword Warning
pcodes P280;P304+P340;P362+P364
MSDS https://store.apolloscientific.co.uk/product/8-chloro-6-trifluoromethyl-124triazolo43-apyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart