APOH30347AFC Apollo Scientific Ltd

4-Chlorobenzofuro[3,2-d]pyrimidine

  • MDL number MFCD00763943
  • Cas Number : 39876-88-5
  • Molecular Weight : 204.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 146
Iupac Name 4-chloro[1]benzofuro[3,2-d]pyrimidine
Inchi Code 1S/C10H5ClN2O/c11-10-9-8(12-5-13-10)6-3-1-2-4-7(6)14-9/h1-5H
Inchi Key ABRHSRPOYMSBOI-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 146
Product Link https://store.apolloscientific.co.uk/product/4-chlorobenzofuro32-dpyrimidine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-chlorobenzofuro32-dpyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart