APOH30347BBD Apollo Scientific Ltd

4-Fluoro-3-cyclopropylphenol

  • MDL number MFCD31700582

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=125269

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=125269
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart