APOH303C39C2 Apollo Scientific Ltd

Methyl 2-bromo-4-methoxybenzoate

  • MDL number MFCD12026001
  • Cas Number : 17100-65-1
  • Molecular Weight : 245.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 25-26
Iupac Name methyl 2-bromo-4-methoxybenzoate
Inchi Code 1S/C9H9BrO3/c1-12-6-3-4-7(8(10)5-6)9(11)13-2/h3-5H,1-2H3
Inchi Key MGIYCRUAYQQSNL-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 25-26
Product Link https://store.apolloscientific.co.uk/product/methyl-2-bromo-4-methoxybenzoate

Safety Information

signalword Danger
pcodes P264;P301+P310
MSDS https://store.apolloscientific.co.uk/product/methyl-2-bromo-4-methoxybenzoate
pictograms GHS06
hstatements H301;H319
Availability details for
Quantity
  • Availability
  • Add to Cart