APOH303C39DA Apollo Scientific Ltd

4,6-Dibromo-2-chloroanisole

  • MDL number MFCD22543700
  • Cas Number : 174913-39-4
  • Molecular Weight : 300.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 75-76
Iupac Name 1,5-dibromo-3-chloro-2-methoxybenzene
density 2.213
Inchi Code 1S/C7H5Br2ClO/c1-11-7-5(9)2-4(8)3-6(7)10/h2-3H,1H3
Inchi Key ITCWWGPAZCJYMO-UHFFFAOYSA-N

Description

Density 2.213
Flash Point 269-272
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 75-76
Product Link https://store.apolloscientific.co.uk/product/46-dibromo-2-chloroanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/46-dibromo-2-chloroanisole
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart