APOH303C39DE Apollo Scientific Ltd

Ethyl 3-amino-6-bromobenzofuran-2-carboxylate

  • MDL number MFCD21609331
  • Cas Number : 887250-14-8
  • Molecular Weight : 284.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-amino-6-bromo-1-benzofuran-2-carboxylate
Inchi Code 1S/C11H10BrNO3/c1-2-15-11(14)10-9(13)7-4-3-6(12)5-8(7)16-10/h3-5H,2,13H2,1H3
Inchi Key OJAYNCRLFMCJIQ-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-3-amino-6-bromobenzofuran-2-carboxylate

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/ethyl-3-amino-6-bromobenzofuran-2-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart